Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqcj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177585.3 | NP_808253.1 | 4 | 361 | ||
GenBank mRNA | AK052136.1 | - | 4 | 362 | ||
BC107305.1 | - | 4 | 325 | 40053239 | ||
BC107306.1 | - | 4 | 325 | 40053241 |
Gene Symbol: Gm21949 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113419.2 | NP_001106890.1 | 4 | 361 | ||
GenBank mRNA | KJ941154.1 | - | 4 | 349 |