Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168525.1 | NP_001161997.1 | 6 | 1649 | ||
NM_001168526.1 | NP_001161998.1 | 2 | 873 | |||
NM_144792.4 | NP_659041.3 | 6 | 1217 | |||
GenBank mRNA | AK076554.1 | - | 4 | 882 | ||
AK079254.1 | - | 2 | 818 | |||
AK082974.1 | - | 6 | 1146 | |||
AK133967.1 | - | 2 | 841 | |||
AK146782.1 | - | 6 | 1217 | |||
AK150272.1 | - | 2 | 850 | |||
AK151170.1 | - | 6 | 1202 | |||
AK152312.1 | - | 2 | 851 | |||
AK153406.1 | - | 6 | 1202 | |||
AK160539.1 | - | 2 | 851 | |||
AY509044.1 | - | 6 | 1217 | |||
BC019443.1 | - | 2 | 840 | 5041121 | ||
BC076618.1 | - | 6 | 1649 | |||
BC085298.1 | - | 3 | 811 | 6307174 |