Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198019.2 | NP_932136.2 | 15 | 2426 | ||
GenBank mRNA | AK160295.1 | - | 7 | 1578 | ||
BC029244.1 | - | 2 | 737 | |||
BC070400.1 | - | 15 | 2426 | 5694341 | ||
BC071270.1 | - | 17 | 2542 |