Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172461.3 | NP_766049.2 | 4, 5 | 697 | ||
GenBank mRNA | AK030042.1 | - | 4, 5 | 656 | ||
AK030186.1 | - | 4, 5 | 700 | |||
AK043543.1 | - | 4, 5 | 936 | |||
AK054237.1 | - | 4, 5 | 591 | |||
BC119588.1 | - | 3, 4 | 531 | 40058889 | ||
BC119589.1 | - | 3, 4 | 531 | 40058890 |