Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145939.2 | NP_666051.2 | 6 | 824 | ||
GenBank mRNA | AK090067.1 | - | 5 | 725 | ||
AK149510.1 | - | 5 | 724 | |||
BC031893.1 | - | 6 | 819 | 5371807 |