Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kntc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042421.1 | NP_001035886.1 | 53 | 5722 | ||
GenBank mRNA | AK084529.1 | - | 10 | 1129 | ||
AK172905.1 | - | 53 | 5696 | |||
BC023686.1 | - | 20 | 2452 |