Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kntc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042421.1 | NP_001035886.1 | 50 | 5411 | ||
GenBank mRNA | AK084529.1 | - | 7 | 818 | ||
AK172905.1 | - | 50 | 5385 | |||
BC023686.1 | - | 17 | 2141 |