Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145363.2 | NP_663338.2 | 2, 3 | 817 | ||
GenBank mRNA | AK084214.1 | - | 2, 3 | 764 | ||
AK148037.1 | - | 2, 3 | 760 | |||
AK148155.1 | - | 3, 4 | 900 | |||
AK148326.1 | - | 2, 3 | 753 | |||
AK148383.1 | - | 2, 3 | 753 | |||
BC019991.1 | - | 2, 3 | 755 | 4189407 |