Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020546.2 | NP_065433.2 | 18 | 2341 | ||
XM_011513942.2 | XP_011512244.1 | 17 | 2150 | |||
GenBank mRNA | AB028983.1 | - | 15 | 1662 | ||
AF070583.1 | - | 2 | 102 | |||
AF410885.1 | - | 18 | 2300 | |||
AK123013.1 | - | 18 | 2188 | |||
AK127036.1 | - | 18 | 2289 | |||
AK294555.1 | - | 15 | 1797 | |||
AK295531.1 | - | 15 | 1681 | |||
AK299775.1 | - | 18 | 1767 | |||
AK304041.1 | - | 18 | 2176 | |||
BC111743.1 | - | 18 | 2279 | 40038765 | ||
DB029098.1 | - | 5 | 465 | |||
L21993.1 | - | 5 | 429 | |||
X74210.1 | - | 5 | 429 |