Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207225.1 | NP_997108.1 | 7 | 878 | ||
GenBank mRNA | AK029933.1 | - | 4 | 415 | ||
AK155250.1 | - | 8 | 1064 | |||
BC066052.1 | - | 7 | 878 | 6827645 |