Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144794.2 | NP_659043.1 | 19 | 1864 | ||
GenBank mRNA | AK049360.1 | - | 19 | 1860 | ||
AK144899.1 | - | 19 | 1917 | |||
AK170559.1 | - | 19 | 1880 | |||
AK173027.1 | - | 19 | 1857 | |||
BC014795.1 | - | 19 | 1837 | 3961992 | ||
BC019442.1 | - | 19 | 1864 | 4036849 |