Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Triqk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171801.1 | NP_001165272.1 | 5 | 926 | ||
NM_173746.3 | NP_776107.2 | 5 | 923 | |||
GenBank mRNA | AK048604.1 | - | 5 | 902 | ||
BC147183.1 | - | 1 | 491 | 9056211 | ||
BC147184.1 | - | 1 | 491 | 9056218 | ||
DQ351292.1 | - | 5 | 924 |