Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpeb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290826.1 | NP_001277755.1 | 2 | 1522 | ||
NM_001290827.1 | NP_001277756.1 | 2 | 1171 | |||
NM_001290828.1 | NP_001277757.1 | 2 | 1233 | |||
NM_001290829.1 | NP_001277758.1 | 2 | 1114 | |||
NM_198300.3 | NP_938042.2 | 2 | 1233 | |||
GenBank mRNA | AB093274.1 | - | 2 | 1162 | ||
AK029261.1 | - | 2 | 1115 | |||
AK044639.1 | - | 2 | 1235 | |||
AK147243.1 | - | 2 | 1523 | |||
AK161513.1 | - | 2 | 1209 | |||
AY313774.1 | - | 2 | 1055 | |||
BC128377.1 | - | 3 | 647 | 40111837 |