Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thnsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001297.2 | NP_001001297.1 | 3 | 685 | ||
NM_177588.2 | NP_808256.1 | 3 | 2257 | |||
GenBank mRNA | AK029518.1 | - | 3 | 2258 | ||
AK033465.1 | - | 3 | 654 | |||
AK083895.1 | - | 3 | 2257 | |||
AK087148.1 | - | 3 | 768 | |||
BC027743.1 | - | 1 | 782 | |||
BC090667.1 | - | 1 | 1966 | 30686917 | ||
BC117854.2 | - | 2 | 2041 | 40094242 |