Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153288.3 | NP_695020.1 | 1 | 226 | ||
GenBank mRNA | AB085945.1 | - | 1 | 194 | ||
AK141698.1 | - | 1 | 227 | |||
BC134361.1 | - | 1 | 198 | 40134626 |