Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pycr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144795.3 | NP_659044.1 | 3, 4 | 755 | ||
GenBank mRNA | AK139185.1 | - | 3, 4 | 756 | ||
BC006727.1 | - | 4, 5 | 400 | 3962093 |