Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tns2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153533.2 | NP_705761.2 | 20, 21 | 3438 | ||
GenBank mRNA | BC025818.1 | - | 3, 4 | 1106 | ||
BC036265.1 | - | 19, 20 | 3369 | |||
BC042190.1 | - | 20, 21 | 3439 | 4188303 |