Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000440.2 | NP_000431.2 | 4 | 893 | ||
XM_017009572.1 | XP_016865061.1 | 2 | 650 | |||
GenBank mRNA | AB593141.1 | - | 2 | 650 | ||
BC035909.1 | - | 4 | 880 | 5492027 | ||
BC144044.1 | - | 4 | 820 | |||
M26061.1 | - | 4 | 893 |