Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271447.1 | NP_001258376.1 | 4 | 461 | ||
NM_001271448.1 | NP_001258377.1 | 3 | 306 | |||
NM_001271449.1 | NP_001258378.1 | 4 | 450 | |||
NM_001271450.1 | NP_001258379.1 | 4 | 450 | |||
NM_001271451.1 | NP_001258380.1 | 4 | 450 | |||
NM_145951.5 | NP_666063.1 | 4 | 405 | |||
GenBank mRNA | AK028343.1 | - | 4 | 451 | ||
AK034058.1 | - | 4 | 406 | |||
AK041822.1 | - | 4 | 451 | |||
AK077508.1 | - | 4 | 432 | |||
AK133956.1 | - | 3 | 277 | |||
AK137736.1 | - | 4 | 432 | |||
AK139897.1 | - | 4 | 432 | |||
BC025915.1 | - | 4 | 424 | 5043650 |