Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145999.1 | NP_001139471.1 | 2 | 533 | ||
NM_001146000.1 | NP_001139472.1 | 2 | 533 | |||
NM_175433.5 | NP_780642.3 | 2 | 616 | |||
GenBank mRNA | AK083979.1 | - | 2 | 458 | ||
AK154871.1 | - | 2 | 514 | |||
AK154953.1 | - | 2 | 534 | |||
AK155416.1 | - | 2 | 595 | |||
BC058640.1 | - | 2 | 427 | 6835601 | ||
BC060641.1 | - | 2 | 510 | 6849964 |