Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027748.3 | NP_082024.2 | 3 | 1011 | ||
GenBank mRNA | AJ292189.1 | - | 3 | 802 | ||
AK017128.1 | - | 1 | 1105 | |||
AK030878.1 | - | 1 | 80 | |||
AK033848.1 | - | 3 | 661 | |||
AK051249.1 | - | 1 | 382 | |||
AK154372.1 | - | 3 | 1020 | |||
BC039989.1 | - | 3 | 967 | |||
BC089030.1 | - | 3 | 850 | |||
BC094546.1 | - | 3 | 968 | |||
BC137617.1 | - | 3 | 952 | 8860636 | ||
BC137618.1 | - | 3 | 952 | 8860637 |