Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpkow | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173747.3 | NP_776108.2 | 6 | 889 | ||
GenBank mRNA | AK028433.1 | - | 6 | 811 | ||
AK144167.1 | - | 6 | 819 | |||
BC062646.1 | - | 4 | 439 | |||
BC092224.1 | - | 6 | 795 | 30355916 | ||
BC120679.1 | - | 5 | 572 | 40129602 | ||
BC125335.1 | - | 5 | 572 | 40129850 |