Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173378.2 | NP_775554.2 | 17 | 3583 | ||
GenBank mRNA | AK133930.1 | - | 2 | 221 | ||
AK134556.1 | - | 10 | 2553 | |||
BC025187.1 | - | 2 | 228 | |||
BC030894.1 | - | 9 | 2212 | |||
BC042874.1 | - | 16 | 3185 |