Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE6G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002602.3 | NP_002593.1 | 4 | 994 | ||
NR_026872.1 | 3 | 789 | ||||
XM_006721942.3 | XP_006722005.1 | 4 | 1967 | |||
XM_017024734.1 | XP_016880223.1 | 4 | 1269 | |||
XM_017024735.1 | XP_016880224.1 | 4 | 1000 | |||
XM_017024736.1 | XP_016880225.1 | 4 | 934 | |||
GenBank mRNA | BQ639341.1 | - | 1 | 522 | ||
BX537414.1 | - | 4 | 1001 | |||
BX647779.1 | - | 3 | 783 | |||
M36476.1 | - | 4 | 950 |