Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007266.1 | NP_001007267.1 | 2 | 814 | ||
GenBank mRNA | AY255589.1 | - | 1 | 310 | ||
AY702326.1 | - | 2 | 814 | |||
BC139118.1 | - | 2 | 867 | 8862140 | ||
BC145814.1 | - | 2 | 867 | 40131117 |
Set Membership: |
G-protein coupled receptor |