Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE6G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002602.3 | NP_002593.1 | 4 | 480 | ||
NR_026872.1 | 3 | 275 | ||||
XM_006721942.3 | XP_006722005.1 | 4 | 1453 | |||
XM_017024734.1 | XP_016880223.1 | 4 | 755 | |||
XM_017024735.1 | XP_016880224.1 | 4 | 486 | |||
XM_017024736.1 | XP_016880225.1 | 4 | 420 | |||
GenBank mRNA | BC106884.2 | - | 4 | 437 | 40029486 | |
BQ638010.1 | - | 4 | 421 | |||
BQ639132.1 | - | 3 | 311 | |||
BX537414.1 | - | 4 | 487 | |||
BX647779.1 | - | 3 | 269 | |||
M36476.1 | - | 4 | 436 |