Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyw3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172474.5 | NP_766062.1 | 5 | 589 | ||
GenBank mRNA | AK030344.1 | - | 5 | 587 | ||
AK034820.1 | - | 5 | 574 | |||
BC086686.1 | - | 5 | 563 | 30680486 | ||
BC087879.1 | - | 5 | 553 | 6533208 | ||
BC141040.1 | - | 5 | 568 | 9055652 |