Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcorl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163073.1 | NP_001156545.1 | 4 | 414 | ||
NM_172153.3 | NP_742165.1 | 4 | 427 | |||
NM_178142.5 | NP_835278.2 | 4 | 414 | |||
GenBank mRNA | AB076078.1 | - | 4 | 426 | ||
AK041987.1 | - | 4 | 373 | |||
AK042567.1 | - | 4 | 428 | |||
AK043498.1 | - | 4 | 372 | |||
AK043845.1 | - | 4 | 382 | |||
AK083689.1 | - | 4 | 376 | |||
AK154398.1 | - | 4 | 375 | |||
AK155421.1 | - | 4 | 375 | |||
BC066151.1 | - | 4 | 344 | |||
BC138563.1 | - | 4 | 414 | 8861583 |