Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166437.1 | NP_001159909.1 | 4 | 503 | ||
NM_172478.3 | NP_766066.2 | 3 | 710 | |||
GenBank mRNA | AK040932.1 | - | 3 | 453 | ||
AK078145.1 | - | 3 | 711 | |||
AK166256.1 | - | 3 | 449 | |||
BC060178.1 | - | 3 | 487 | |||
BC113777.1 | - | 4 | 451 | |||
BC128565.1 | - | 4 | 453 | |||
BC128566.1 | - | 3 | 660 | 40094900 |