Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243137.1 | NP_001230066.1 | 20 | 2136 | ||
NM_002605.2 | NP_002596.1 | 20 | 2338 | |||
NM_173454.1 | NP_775656.1 | 19 | 2200 | |||
XM_011521668.2 | XP_011519970.1 | 13 | 2472 | |||
XM_017022308.1 | XP_016877797.1 | 17 | 2569 | |||
XM_017022309.1 | XP_016877798.1 | 18 | 1936 | |||
XM_017022311.1 | XP_016877800.1 | 21 | 3073 | |||
GenBank mRNA | AF056490.1 | - | 18 | 1801 | ||
AF332653.1 | - | 20 | 2285 | |||
AF388183.1 | - | 20 | 2331 | |||
AF388184.1 | - | 19 | 2193 | |||
AF388185.1 | - | 18 | 2114 | |||
AF388186.1 | - | 19 | 2252 | |||
AF388187.1 | - | 21 | 2529 | |||
AK074280.1 | - | 20 | 2136 | |||
AK122689.1 | - | 3 | 1021 | |||
AK127232.1 | - | 17 | 3619 | |||
AL109687.1 | - | 2 | 142 | |||
AL109778.1 | - | 2 | 197 | |||
BC013825.1 | - | 3 | 251 | |||
BC037260.1 | - | 3 | 251 | |||
BC048317.1 | - | 4 | 578 | |||
BC060762.1 | - | 21 | 2401 | 30344110 | ||
BC075822.1 | - | 20 | 2229 | 6296489 |