Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113211.1 | NP_001106682.1 | 7 | 802 | ||
NM_173732.3 | NP_776093.1 | 7 | 802 | |||
GenBank mRNA | AK090031.1 | - | 7 | 781 | ||
AK129110.1 | - | 7 | 799 | |||
BC030440.1 | - | 7 | 813 | 5373527 | ||
DQ695550.1 | - | 1 | 18 |