Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243137.1 | NP_001230066.1 | 10, 11 | 988 | ||
NM_002605.2 | NP_002596.1 | 10, 11 | 1190 | |||
NM_173454.1 | NP_775656.1 | 9, 10 | 1052 | |||
XM_011521668.2 | XP_011519970.1 | 3, 4 | 1324 | |||
XM_017022308.1 | XP_016877797.1 | 7, 8 | 1421 | |||
XM_017022309.1 | XP_016877798.1 | 8, 9 | 788 | |||
XM_017022310.1 | XP_016877799.1 | 10, 11 | 1727 | |||
XM_017022311.1 | XP_016877800.1 | 11, 12 | 1925 | |||
GenBank mRNA | AF056490.1 | - | 8, 9 | 653 | ||
AF332653.1 | - | 10, 11 | 1137 | |||
AF388183.1 | - | 10, 11 | 1183 | |||
AF388184.1 | - | 9, 10 | 1045 | |||
AF388185.1 | - | 8, 9 | 966 | |||
AF388186.1 | - | 9, 10 | 1104 | |||
AF388187.1 | - | 11, 12 | 1381 | |||
AK074280.1 | - | 10, 11 | 988 | |||
AK127232.1 | - | 7, 8 | 2471 | |||
BC060762.1 | - | 11, 12 | 1253 | 30344110 | ||
BC075822.1 | - | 10, 11 | 1081 | 6296489 |