Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252478.1 | NP_001239407.1 | 8 | 533 | ||
NM_144798.6 | NP_659047.2 | 8 | 518 | |||
GenBank mRNA | AF233346.1 | - | 7 | 470 | ||
AF395840.1 | - | 7 | 460 | |||
AK082907.1 | - | 8 | 495 | |||
AK150342.1 | - | 8 | 494 | |||
AK151120.1 | - | 8 | 494 | |||
AK160903.1 | - | 8 | 522 | |||
BC066162.1 | - | 8 | 485 | 30471027 | ||
BC068169.1 | - | 8 | 507 | 30542277 |