Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE9A SIRNA Assays ›
Gene Symbol:
PDE9A
Gene Name:
phosphodiesterase 9A
Gene Aliases:
HSPDE9A2
Chromosome Location:
Chr.21: 42653752 - 42775509 on Build GRCh38
UniGene ID:
Hs.473927
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s10224
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE9A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001567.1
NP_001001567.1
15
1323
NM_001001568.1
NP_001001568.1
13
1174
NM_001001569.1
NP_001001569.1
12
1119
NM_001001570.1
NP_001001570.1
15
1344
NM_001001571.1
NP_001001571.1
14
1252
NM_001001572.1
NP_001001572.1
12
1130
NM_001001573.1
NP_001001573.1
13
1310
NM_001001574.1
NP_001001574.1
14
1245
NM_001001575.1
NP_001001575.1
11
1075
NM_001001576.1
NP_001001576.1
11
1034
NM_001001577.1
NP_001001577.1
12
1153
NM_001001578.1
NP_001001578.1
13
1197
NM_001001579.1
NP_001001579.1
13
1190
NM_001001580.1
NP_001001580.1
10
990
NM_001001581.1
NP_001001581.1
14
1354
NM_001001582.1
NP_001001582.1
15
1432
NM_001001583.1
NP_001001583.1
15
1425
NM_001001584.2
NP_001001584.1
14
1253
NM_001001585.1
NP_001001585.1
14
1377
NM_001315533.1
NP_001302462.1
16
1399
NM_002606.2
NP_002597.1
16
1503
XM_011529598.2
XP_011527900.1
16
1425
XM_017028366.1
XP_016883855.1
14
1241
GenBank mRNA
AF048837.1
-
16
1500
AF067223.1
-
16
1452
AF067224.1
-
15
1271
AF067225.1
-
13
1113
AF067226.1
-
12
1058
AK001165.1
-
13
1105
AK001307.1
-
15
1294
AK127770.1
-
6
2854
AK294112.1
-
13
1140
AK314679.1
-
16
1476
AY196299.1
-
15
1317
AY196300.1
-
14
1225
AY196301.1
-
12
1103
AY196302.1
-
13
1283
AY196303.1
-
14
1218
AY196304.1
-
11
1048
AY196305.1
-
11
1007
AY196306.1
-
12
1126
AY196307.1
-
13
1170
AY196308.1
-
13
1163
AY196309.1
-
10
963
AY196310.1
-
14
1327
AY196311.1
-
15
1405
AY196312.1
-
15
1398
AY196313.1
-
14
1225
AY196314.1
-
14
1350
AY242121.1
-
14
1189
AY701187.1
-
16
1396
BC009047.1
-
15
1297
3874635
BT007016.1
-
15
1260
KU178246.1
-
15
1260
KU178247.1
-
16
1440
KU178248.1
-
12
1056
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphodiesterase
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
positive regulation of cardiac muscle hypertrophy
cGMP-mediated signaling
cGMP metabolic process
cGMP catabolic process
3',5'-cyclic-nucleotide phosphodiesterase activity
protein binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline