Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201638.2 | NP_964000.2 | 9 | 986 | ||
GenBank mRNA | AK129409.1 | - | 944 | |||
AK146881.1 | - | 9 | 975 | |||
AK153810.1 | - | 9 | 982 | |||
BC044899.1 | - | 1 | 672 | |||
BC052204.1 | - | 9 | 944 | 30007932 |