Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181584.4 | NP_853615.1 | 9 | 1809 | ||
GenBank mRNA | AJ748654.1 | - | 6 | 656 | ||
AK031227.1 | - | 9 | 1807 | |||
AY057988.1 | - | 9 | 1695 | |||
BC141240.1 | - | 9 | 1692 | 9055852 |