Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcmbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145955.3 | NP_666067.1 | 15, 16 | 2045 | ||
GenBank mRNA | AK196573.1 | - | 2, 3 | 153 | ||
BC025641.1 | - | 15, 16 | 2075 | 5322565 | ||
BC038342.1 | - | 15, 16 | 2045 | 4191750 |