Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199251.1 | NP_954859.1 | 2 | 1792 | ||
GenBank mRNA | AB052939.1 | - | 2 | 1792 | ||
BC138133.1 | - | 2 | 1584 | 8861152 | ||
BC138134.1 | - | 2 | 1584 | 8861153 |