Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153135.3 | NP_694775.1 | 3, 4 | 758 | ||
GenBank mRNA | AJ487854.1 | - | 3, 4 | 759 | ||
BC137615.1 | - | 3, 4 | 549 | 8860634 | ||
BC144783.1 | - | 3, 4 | 549 | 9053331 | ||
BC144784.1 | - | 3, 4 | 549 | 9053332 | ||
BC145695.1 | - | 3, 4 | 549 | 40130998 | ||
FJ824810.1 | - | 3, 4 | 691 |