Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253731.1 | NP_001240660.1 | 14 | 1808 | ||
NM_153414.4 | NP_700463.2 | 14 | 1776 | |||
GenBank mRNA | AK038979.1 | - | 14 | 1509 | ||
AK077634.1 | - | 14 | 1777 | |||
AK136992.1 | - | 14 | 1503 | |||
AK150195.1 | - | 14 | 1503 | |||
BC028953.1 | - | 13 | 1466 | 5362016 | ||
BC055700.1 | - | 14 | 1475 | 6808816 |