Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253731.1 | NP_001240660.1 | 5 | 638 | ||
NM_153414.4 | NP_700463.2 | 5 | 606 | |||
GenBank mRNA | AK035769.1 | - | 5 | 350 | ||
AK038979.1 | - | 5 | 339 | |||
AK077634.1 | - | 5 | 607 | |||
AK136992.1 | - | 5 | 333 | |||
AK150195.1 | - | 5 | 333 | |||
BC028953.1 | - | 4 | 296 | 5362016 | ||
BC055700.1 | - | 5 | 305 | 6808816 |