Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrtm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172207.1 | NP_001165678.1 | 5 | 1200 | ||
NM_172492.3 | NP_766080.1 | 5 | 1371 | |||
GenBank mRNA | AK039006.1 | - | 5 | 1205 | ||
AK045494.1 | - | 5 | 1285 | |||
BC058944.1 | - | 4 | 1761 | 6827376 |