Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170673.3 | NP_733773.2 | 12, 13 | 933 | ||
GenBank mRNA | AK042692.1 | - | 12, 13 | 934 | ||
AK044080.1 | - | 12, 13 | 899 | |||
AK047571.1 | - | 12, 13 | 877 | |||
AK137422.1 | - | 12, 13 | 934 | |||
AK220550.1 | - | 5, 6 | 1870 | |||
BC115728.1 | - | 12, 13 | 883 | 40039295 |