Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000283.3 | NP_000274.2 | 19 | 2304 | ||
NM_001145291.1 | NP_001138763.1 | 19 | 2304 | |||
NM_001145292.1 | NP_001138764.1 | 17 | 1683 | |||
XM_011513473.2 | XP_011511775.1 | 20 | 3571 | |||
XM_011513474.2 | XP_011511776.1 | 20 | 3571 | |||
XM_011513475.2 | XP_011511777.1 | 22 | 3674 | |||
XM_011513476.2 | XP_011511778.1 | 20 | 3571 | |||
XM_011513477.2 | XP_011511779.1 | 15 | 2423 | |||
XM_011513478.2 | XP_011511780.1 | 16 | 2490 | |||
XM_017008283.1 | XP_016863772.1 | 17 | 1710 | |||
XM_017008284.1 | XP_016863773.1 | 17 | 1665 | |||
XM_017008285.1 | XP_016863774.1 | 17 | 1836 | |||
XM_017008286.1 | XP_016863775.1 | 17 | 1665 | |||
XM_017008287.1 | XP_016863776.1 | 16 | 1766 | |||
XM_017008288.1 | XP_016863777.1 | 14 | 2375 | |||
XM_017008289.1 | XP_016863778.1 | 15 | 1563 | |||
GenBank mRNA | AK295285.1 | - | 17 | 1680 | ||
AK316054.1 | - | 17 | 1640 | |||
BC000249.1 | - | 19 | 2294 | 3356353 | ||
BT009794.1 | - | 19 | 2251 | |||
S41458.1 | - | 19 | 2272 | |||
X66142.1 | - | 19 | 2256 |