Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178591.2 | NP_848706.2 | 1 | 233 | ||
GenBank mRNA | AK048708.1 | - | 1 | 415 | ||
AK141037.1 | - | 1 | 421 | |||
AK164499.1 | - | 1 | 387 | |||
AY648975.1 | - | 1 | 233 | |||
BC151113.1 | - | 1 | 317 | 9088026 |