Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178591.2 | NP_848706.2 | 6 | 1041 | ||
GenBank mRNA | AK141037.1 | - | 6 | 1229 | ||
AK170314.1 | - | 6 | 549 | |||
AY648975.1 | - | 6 | 1041 | |||
AY648976.1 | - | 10 | 876 | |||
BC151113.1 | - | 6 | 1125 | 9088026 |