Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145962.2 | NP_666074.1 | 2, 3 | 803 | ||
GenBank mRNA | AK039865.1 | - | 2, 3 | 805 | ||
AK136305.1 | - | 2, 3 | 493 | |||
AK163377.1 | - | 1, 2 | 190 | |||
BC027089.1 | - | 2, 3 | 415 | 5355443 | ||
BC032188.1 | - | 2, 3 | 849 |