Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem87a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110496.1 | NP_001103966.1 | 9 | 919 | ||
NM_001110497.1 | NP_001103967.1 | 9 | 919 | |||
NM_173734.3 | NP_776095.2 | 9 | 919 | |||
GenBank mRNA | AK044591.1 | - | 9 | 873 | ||
AK086738.1 | - | 9 | 869 | |||
AK088559.1 | - | 9 | 901 | |||
AK134268.1 | - | 9 | 882 | |||
BC004047.1 | - | 9 | 868 | |||
BC027354.1 | - | 9 | 892 | 4010477 |