Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005390.4 | NP_005381.1 | 1 | 1029 | ||
GenBank mRNA | AK313872.1 | - | 1 | 989 | ||
BC030697.2 | - | 1 | 1025 | 4824350 | ||
BC066953.1 | - | 1 | 1011 | 5273609 | ||
BC094760.1 | - | 1 | 1024 | |||
BC119656.1 | - | 1 | 997 | 40115940 | ||
BC119657.1 | - | 1 | 997 | 40115941 | ||
BC127637.1 | - | 1 | 997 | 40115943 | ||
BC127638.1 | - | 1 | 996 | 40115944 |