Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004364.2 | NP_001004364.2 | 6 | 810 | ||
NM_001098168.1 | NP_001091637.1 | 6 | 810 | |||
NM_001135192.1 | NP_001128664.1 | 6 | 810 | |||
GenBank mRNA | AK147622.1 | - | 6 | 818 | ||
AK162242.1 | - | 109 | ||||
AK172944.1 | - | 6 | 622 | |||
BC080847.1 | - | 6 | 810 | 6852788 | ||
BC096022.1 | - | 6 | 578 | 30542525 |